Order Entry
Ireland
ContactUsLinkComponent
L(-)-Tryptophan, Millipore®
L(-)-Tryptophan, Millipore®
Catalog # 6540-100
CAS Number:  
L(-)-Tryptophan, Millipore®
Catalog # 6540-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :204.23 g/mol
Merck Index:12,09929
Melting Pt:290 °C
Density:1,34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-795-6
CAS Number:73-22-3
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00064340