L(+)-Potassium sodium tartrate tetrahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:6381-59-5
:
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989
:282.22 g/mol
Merck Index:13,07755
Melting Pt:70…80 °C
Density:∼1,767 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-156-8
Frequently Bought Together