D-(-)-Tartaric acid
Catalog # ROTH5529.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:150.09 g/mol
Merck Index:12,09235
Boiling Pt:356 °C (1013 hPa)
Melting Pt:166…169 °C
Density:1,76 g/cm³ (20 °C)
Flash Pt:210 °C
Storage Temperature:Ambient
EINECS:205-695-6
:GHS07
CAS Number:147-71-7
Formula:C₄H₆O₆
MDL Number:MFCD00004238