Order Entry
Ireland
ContactUsLinkComponent
D-(-)-Tartaric acid
D-(-)-Tartaric acid
Catalog # MOLE17155420-100G
Supplier:  Molekula
CAS Number:  
D-(-)-Tartaric acid
Catalog # MOLE17155420-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :150.09 g/mol
Merck Index:12,09235
Boiling Pt:356 °C (1013 hPa)
Melting Pt:166…169 °C
Density:1,76 g/cm³ (20 °C)
Flash Pt:210 °C
Storage Temperature:Ambient
EINECS:205-695-6
 :GHS07
CAS Number:147-71-7
Formula:C₄H₆O₆
MDL Number:MFCD00004238