Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, α-Hexabromocyclododecane 50 µg/ml in toluene, CIL
Organic reference standard, α-Hexabromocyclododecane 50 µg/ml in toluene, CIL
  PROOCILULM48341.2
 :  
Organic reference standard, α-Hexabromocyclododecane 50 µg/ml in toluene, CIL
  PROOCILULM48341.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    134237-50-6
  • Description:
    α-Hexabromocyclododecane 50 µg/ml in toluene
  • Formula:
    C₁₂H₁₈Br₆
  • Description:
    α-Hexabromocyclododecane