Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, β-Hexabromocyclododecane 50 µg/ml in toluene, CIL
  PROOCILULM48351.2
 :  
Organic reference standard, β-Hexabromocyclododecane 50 µg/ml in toluene, CIL
  PROOCILULM48351.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    134237-51-7
  • Description:
    β-Hexabromocyclododecane 50 µg/ml in toluene
  • Formula:
    C₁₂H₁₈Br₆
  • Description:
    β-Hexabromocyclododecane