Organic reference standard, Decachlorobiphenyl (PCB No. 209) 35 µg/ml in isooctane
Catalog # PROOCIL-PCB-209
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2051-24-3
- Description:Decachlorobiphenyl (PCB No. 209) 35 µg/ml in isooctane
- Formula:C₁₂Cl₁₀
- Description:Decachlorobiphenyl (PCB No. 209)