Organic reference standard, 2,4,6-Trichlorobiphenyl (PCB No. 30) 100 µg/ml in isooctane, CIL
PROOCIL-PCB-30-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:35693-92-6
- Description:2,4,6-Trichlorobiphenyl (PCB No. 30) 100 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,4,6-Trichlorobiphenyl (PCB No. 30)