Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 4,4'-Dichlorobiphenyl (PCB No. 15) 35 µg/ml in isooctane, CIL
Organic reference standard, 4,4'-Dichlorobiphenyl (PCB No. 15) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-15
CAS Number:  
Organic reference standard, 4,4'-Dichlorobiphenyl (PCB No. 15) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-15
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2050-68-2
  • Description:
    4,4'-Dichlorobiphenyl (PCB No. 15) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₈Cl₂
  • Description:
    4,4'-Dichlorobiphenyl (PCB No. 15)

Specifications

About this item