Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2-Chlorobiphenyl (PCB No. 1) 35 µg/ml in isooctane, CIL
Organic reference standard, 2-Chlorobiphenyl (PCB No. 1) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-1
CAS Number:  
Organic reference standard, 2-Chlorobiphenyl (PCB No. 1) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2051-60-7
  • Description:
    2-Chlorobiphenyl (PCB No. 1) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₉Cl
  • Description:
    2-Chlorobiphenyl (PCB No. 1)

Specifications

About this item