Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,2'-Dichlorobiphenyl (PCB No. 4) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,2'-Dichlorobiphenyl (PCB No. 4) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-4-CS
 :  
Organic reference standard, 2,2'-Dichlorobiphenyl (PCB No. 4) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-4-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    13029-08-8
  • Description:
    2,2'-Dichlorobiphenyl (PCB No. 4) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₈Cl₂
  • Description:
    2,2'-Dichlorobiphenyl (PCB No. 4)