Organic reference standard, 3,4,4'-Trichlorobiphenyl (PCB No. 37) 100 µg/ml in isooctane, CIL
PROOCIL-PCB-37-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:38444-90-5
- Description:3,4,4'-Trichlorobiphenyl (PCB No. 37) 100 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:3,4,4'-Trichlorobiphenyl (PCB No. 37)