Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 3,4,4',5-Tetrachlorobiphenyl (PCB No. 81) 35 µg/ml in isooctane, CIL
Organic reference standard, 3,4,4',5-Tetrachlorobiphenyl (PCB No. 81) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-81
CAS Number:  
Organic reference standard, 3,4,4',5-Tetrachlorobiphenyl (PCB No. 81) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-81
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    70362-50-4
  • Description:
    3,4,4',5-Tetrachlorobiphenyl (PCB No. 81) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    3,4,4',5-Tetrachlorobiphenyl (PCB No. 81)

Specifications

About this item