Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,4',6-Trichlorobiphenyl (PCB No. 32) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,4',6-Trichlorobiphenyl (PCB No. 32) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-32-CS
CAS Number:  
Organic reference standard, 2,4',6-Trichlorobiphenyl (PCB No. 32) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-32-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    38444-77-8
  • Description:
    2,4',6-Trichlorobiphenyl (PCB No. 32) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,4',6-Trichlorobiphenyl (PCB No. 32)

Specifications

About this item