Organic reference standard, 2,3,4,4',6-Pentachlorobiphenyl (PCB No. 115) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-115
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-38-1
- Description:2,3,4,4',6-Pentachlorobiphenyl (PCB No. 115) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,4,4',6-Pentachlorobiphenyl (PCB No. 115)