Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-110-CS
 :  
Organic reference standard, 2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-110-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    38380-03-9
  • Description:
    2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)