Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-105
CAS Number:  
Organic reference standard, 2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-105
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    32598-14-4
  • Description:
    2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105)

Specifications

About this item