Organic reference standard, 2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-105-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:32598-14-4
- Description:2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105) 100 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105)