Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, 2,2',6-Trichlorobiphenyl (PCB No. 19) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-19-CS
 :  
Organic reference standard, 2,2',6-Trichlorobiphenyl (PCB No. 19) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-19-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    38444-73-4
  • Description:
    2,2',6-Trichlorobiphenyl (PCB No. 19) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,2',6-Trichlorobiphenyl (PCB No. 19)