Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,2',3-Trichlorobiphenyl (PCB No. 16) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3-Trichlorobiphenyl (PCB No. 16) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-16
 :  
Organic reference standard, 2,2',3-Trichlorobiphenyl (PCB No. 16) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-16
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    38444-78-9
  • Description:
    2,2',3-Trichlorobiphenyl (PCB No. 16) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,2',3-Trichlorobiphenyl (PCB No. 16)