Organic reference standard, 2,2',4,4',5-Pentachlorobiphenyl (PCB No. 99) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-99
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:38380-01-7
- Description:2,2',4,4',5-Pentachlorobiphenyl (PCB No. 99) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',4,4',5-Pentachlorobiphenyl (PCB No. 99)