Organic reference standard, 2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-53
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:41464-41-9
- Description:2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',5,6'-Tetrachlorobiphenyl (PCB No. 53)