Organic reference standard, 3,5-Dichlorobiphenyl (PCB No. 14) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-14
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:34883-41-5
- Description:3,5-Dichlorobiphenyl (PCB No. 14) 35 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:3,5-Dichlorobiphenyl (PCB No. 14)