Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, 2,3',5',6-Tetrachlorobiphenyl (PCB No. 73) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-73
CAS Number:  
Organic reference standard, 2,3',5',6-Tetrachlorobiphenyl (PCB No. 73) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-73
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    74338-23-1
  • Description:
    2,3',5',6-Tetrachlorobiphenyl (PCB No. 73) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,3',5',6-Tetrachlorobiphenyl (PCB No. 73)

Specifications

About this item