Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,2',6,6'-Tetrachlorobiphenyl (PCB No. 54) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,2',6,6'-Tetrachlorobiphenyl (PCB No. 54) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-54-CS
 :  
Organic reference standard, 2,2',6,6'-Tetrachlorobiphenyl (PCB No. 54) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-54-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    15968-05-5
  • Description:
    2,2',6,6'-Tetrachlorobiphenyl (PCB No. 54) 100 µg/ml in isooctane
  • Environmentally Preferable:
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,2',6,6'-Tetrachlorobiphenyl (PCB No. 54)