Organic reference standard, 2,3,3',4',5',6-Hexachlorobiphenyl (PCB No. 164) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-164
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-45-0
- Description:2,3,3',4',5',6-Hexachlorobiphenyl (PCB No. 164) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3,3',4',5',6-Hexachlorobiphenyl (PCB No. 164)