Organic reference standard, 3,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 169) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-169
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:32774-16-6
- Description:3,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 169) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:3,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 169)