Organic reference standard, 2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-157-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:69782-90-7
- Description:2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157) 100 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)