Organic reference standard, 2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-138
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:35065-28-2
- Description:2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)