Organic reference standard, 2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-160
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:41411-62-5
- Description:2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3,3',4,5,6-Hexachlorobiphenyl (PCB No. 160)