Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, p-Xylene 200 µg/ml in methanol
Organic reference standard, p-Xylene 200 µg/ml in methanol
123372H
:
AccuStandard
:
Organic reference standard, p-Xylene 200 µg/ml in methanol
123372H
:
AccuStandard
:
Pack type:
Glass bottle
Pk:
1 mL
CAS No.:
106-42-3
Description:
p-Xylene 200 µg/ml in methanol
Formula:
C₈H₁₀
Description:
p-Xylene