Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 1,2-Dichloroethane 5000 µg/ml in methanol
Organic reference standard, 1,2-Dichloroethane 5000 µg/ml in methanol
Catalog # PROONE5612
CAS Number:  
Organic reference standard, 1,2-Dichloroethane 5000 µg/ml in methanol
Catalog # PROONE5612
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • CAS No.:
    107-06-2
  • Description:
    1,2-Dichloroethane 5,000 µg/ml in methanol
  • Formula:
    C₂H₄Cl₂
  • Description:
    1,2-Dichloroethane

Specifications

About this item