Order Entry
Ireland
ContactUsLinkComponent
Cobalt(II) chloride hexahydrate 98-102%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Cobalt(II) chloride hexahydrate 98-102%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Catalog # 31277-100G
CAS Number:  
Cobalt(II) chloride hexahydrate 98-102%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Catalog # 31277-100G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.005 %
  • Potassium (K):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.002 %
  • Magnesium (Mg):
    Max. 0.001 %
  • Manganese (Mn):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.01 %
  • Nickel (Ni):
    Max. 0.05 %
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.005 %
  • Nitrate (NO₃):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :237.93 g/mol
Merck Index:13,02462
Boiling Pt:Decomposes on heating
Boiling Pt: Dec. (on heating)
Melting Pt:86 °C
Density:1,9 g/cm³ (20 °C)
EINECS:231-589-4
ADR:9,III
 :GHS07
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652