Cobalt(II) chloride hexahydrate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-13-1
:
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652
:237.93 g/mol
Merck Index:14,2437
Boiling Pt:110 °C (1013 hPa)
Melting Pt:87 °C
Density:1,924
Storage Temperature:Ambient
EINECS:231-589-4
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Related Information
Frequently Bought Together