Order Entry
Ireland
ContactUsLinkComponent
Zinc sulphate heptahydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, SAFC®
Corrosive
Irritant
Environment
Zinc sulphate heptahydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, SAFC®
Catalog # 1.08881.1000
CAS Number:  
Corrosive
Irritant
Environment
Zinc sulphate heptahydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, FCC, SAFC®
Catalog # 1.08881.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:12,10293
Melting Pt:100 °C
Density:1,97 g/cm³ (20 °C)
ADR:9,III
 :287.54 g/mol
EINECS:231-793-3
UN:3077
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:7446-20-0
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894