Order Entry
Ireland
ContactUsLinkComponent
 
 
 
 
Copper(II) nitrate trihydrate 99-104%, Puriss. p.a., Fluka™
 
 
 
 
Copper(II) nitrate trihydrate 99-104%, Puriss. p.a., Fluka™
 

 

  • Assay:
    99.0 - 104.0 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Iron (Fe):
    Max. 20 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 10 ppm
  • Lead (Pb):
    Max. 10 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 50 ppm
  • Chloride (Cl):
    Max. 20 ppm

 

 

Frequently Bought Together