


Copper(II) nitrate trihydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:10031-43-3
:
Formula:Cu(NO₃)₂·3H₂O
MDL Number:MFCD00149671
:241.6 g/mol
Merck Index:13,02671
Melting Pt:114 °C
Density:2,05 g/cm³ (20 °C)
EINECS:221-838-5
UN:1477
ADR:5.1,II
:GHS03
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together