Order Entry
Ireland
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
Ammonium heptamolybdate tetrahydrate ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates
Ammonium heptamolybdate tetrahydrate ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    400 g/l (20 °C)
  • Melting Point:
    90 °C Elimination of water of crystallisation
  • Molar Mass:
    1235.86 g/mol
  • Bulk Density:
    800 kg/m3
  • Density:
    2.498 g/cm3 (20 °C)
  • pH:
    5.3 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together