Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tin (II) chloride dihydrate
Tin(II) chloride dihydrate
Tin(II) chloride dihydrate
:
Roth Carl
Tin(II) chloride dihydrate
:
Roth Carl
:
10025-69-1
:
:
SnCl₂·2H₂O
:
MFCD00149863
:
225.65 g/mol
:
13,08861
:
623 °C (1013 hPa)
:
246 °C
:
2,71 g/cm³ (25 °C)
:
Ambient
:
231-868-0
:
3260
:
8,III
:
GHS05
:
GHS07
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetic anhydride for synthesis, Sigma-Aldrich®
: Merck
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
: VWR Chemicals
Ethanol absolute, BAKER ANALYZED® HPLC for HPLC, analytical reagent, J.T.Baker®
: Avantor