

Magnesium perchlorate hydrate 1-4 mm drying agent, Supelco®
Catalog # 1.05873.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Metal can
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:241.22 g/mol
Melting Pt:146 °C
Density:2,6 g/cm³ (25 °C) (anhydr.)
ADR:5.1,II
:M-Clarity™ quality level = MQ200
EINECS:233-108-3
UN:1475
:GHS03
:GHS07
Storage Temperature:Ambient
CAS Number:64010-42-0
Formula:Cl₂MgO₈·H₂O