Order Entry
Ireland
Orders LinkContactUsLinkComponent
Calcium bromide hydrate for analysis

Calcium bromide hydrate for analysis

Corrosive
Calcium bromide hydrate for analysis
Certificates
Calcium bromide hydrate for analysis

3 Options Available Below

Product Details & Documents
  • CAS Number:71626-99-8
  • Formula:Br₂Ca·H₂O
  • MDL Number:MFCD00149608
  • Molecular Weight:217.9 g/mol
  • Melting Pt:730 °C
  • Density:2,295 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-164-6

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder and/or chunks
  • Titration Complexometric:
    ≥83.0 % (CaBr2)
  • Acidity/alkalinity:
    Passes test
  • Chloride (Cl):
    ≤2000 ppm
  • Sulfate (SO4):
    ≤100 ppm
  • Bromate and Chlorate:
    Passes test
  • Arsenic (As):
    ≤2 ppm
  • Barium (Ba):
    Passes test
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Subst not pptd:
    ≤0.4 % By ammonium oxalate (as SO4)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 189,00 €
    189,00 €
    1 * 500 g
       189,00 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 334,00 €
    334,00 €
    1 * 1 kg
       334,00 €

  • Plastic bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 674,00 €
    674,00 €
    1 * 2,5 kg
       674,00 €