Sodium dihydrogen phosphate dihydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:13472-35-0
:
Formula:NaH₂PO₄·2H₂O
MDL Number:MFCD00149209
:156.01 g/mol
Merck Index:12,08806
Boiling Pt:∼650 °C (1013 hPa)
Melting Pt:60 °C
Density:1,91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Frequently Bought Together