Order Entry
Ireland
ContactUsLinkComponent
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Ba (Barium):
    Passes test
  • Insolubility in water:
    Max. 0.03 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 50 ppm
  • Na (Sodium):
    Max. 0.02 %

Specification Test Results

Frequently Bought Together