

tetra-Chloroauric(III) acid hydrate ≥99.999% (metals basis) Au ≥ 49.0%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27988-77-8
:
Formula:HAuCl₄·H₂O
MDL Number:MFCD00149903
:357.8 g/mol
Density:1,169 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-948-4
UN:3260
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together