Order Entry
Ireland
ContactUsLinkComponent
1-Octanesulphonic acid sodium salt, hydrate 99+% for HPLC
1-Octanesulphonic acid sodium salt, hydrate 99+% for HPLC
  • Loss on drying:
    6.9-8.5%
  • 10% water solutionClear:
    Pass Test.
  • colorless 0.4% EtOH/MeOHClear:
    Pass Test
  • clear0.4% EtOH/MeOH:
    Clear
  • IR identification:
    Pass test
Frequently Bought Together