di-Ammonium oxalate monohydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:6009-70-7
:
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694
:142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:∼1,5 g/cm³ (20 °C)
UN:3288
ADR:6.1,III
:M-Clarity™ quality level = MQ300
EINECS:214-202-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:45 g/l (20 °C) (anhydrous substance)
- Melting Point:70 °C (decomposition)
- Molar Mass:142.11 g/mol
- Bulk Density:480 kg/m3
- Vapor Pressure:(20 °C)
- Density:1.50 g/cm3 (20 °C)
- pH:6.3 (50 g/l, H2O, 25 °C)
Specification Test Results
Related Information
Frequently Bought Together