Cobalt(II) acetate tetrahydrate (24% Co)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6147-53-1
:
Formula:(H₃CCOO)₂Co·4H₂O
MDL Number:MFCD00149569
:249.08 g/mol
Merck Index:14,2433
Melting Pt:∼140 °C (-4H₂O)
Density:1,71
Storage Temperature:Ambient
EINECS:200-755-8
UN:3077
ADR:9,III
:GHS08
:GHS07
Related Information
Frequently Bought Together