Order Entry
Ireland
ContactUsLinkComponent
 
Health
Toxic
Environment
Hydrazinium sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 24696.186
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Toxic
Environment
Hydrazinium sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 24696.186
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue:
    Max. 0.05 %
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :130.12 g/mol
Merck Index:14,04772
Melting Pt:254 °C
Density:1,37 g/cm³ (20 °C)
EINECS:233-110-4
UN:3288
ADR:6.1,III
 :GHS08
 :GHS06
 :GHS09
CAS Number:10034-93-2
Formula:NH₂NH₂·H₂SO₄
MDL Number:MFCD00044873