Order Entry
Ireland
ContactUsLinkComponent
Guanidinium hydrochloride
Irritant
Guanidinium hydrochloride
Catalog # A1106.1000
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride
Catalog # A1106.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1,354 g/cm³ (20 °C)
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026