Order Entry
Ireland
ContactUsLinkComponent
Spermidine trihydrochloride
Spermidine trihydrochloride
Catalog # MOLE31867014-5G
Supplier:  Molekula
CAS Number:  
Spermidine trihydrochloride
Catalog # MOLE31867014-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :254.63 g/mol
Merck Index:12,08893
Melting Pt:256…258 °C
Storage Temperature:Freezer
EINECS:206-379-0
CAS Number:334-50-9
Formula:C₇H₂₂Cl₃N₃
MDL Number:MFCD00012918